Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oc90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010953.2 | NP_035083.1 | 8 - 9 | 718 | 59 | |
GenBank mRNA | AF091846.1 | - | 6 - 7 | 572 | 59 | |
AF093591.1 | - | 8 - 9 | 828 | 59 | ||
AK012981.1 | - | 8 - 9 | 720 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |