Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290453.1 | NP_001277382.1 | 3 - 4 | 906 | 65 | |
NM_145819.2 | NP_665818.2 | 3 - 4 | 906 | 65 | ||
GenBank mRNA | AB007407.1 | - | 2 - 3 | 1125 | 65 | |
AB088478.1 | - | 3 - 4 | 807 | 65 | ||
AK030410.1 | - | 3 - 4 | 907 | 65 | ||
AK158837.1 | - | 1 - 2 | 2158 | 65 | ||
BC060649.1 | - | 3 - 4 | 866 | 65 | ||
BC129905.1 | - | 3 - 4 | 656 | 40111655 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |