Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012312.1 | - | 1 - 2 | 175 | 76 | |
AK016651.1 | - | 1 - 2 | 318 | 76 | ||
AK018787.1 | - | 1 - 2 | 185 | 76 | ||
AK038578.1 | - | 1 - 2 | 139 | 76 | ||
AK044291.1 | - | 1 - 2 | 288 | 76 | ||
AK048044.1 | - | 1 - 2 | 288 | 76 | ||
AK076231.1 | - | 1 - 2 | 302 | 76 | ||
AK078139.1 | - | 1 - 2 | 286 | 76 | ||
AK078468.1 | - | 1 - 2 | 302 | 76 | ||
AK078872.1 | - | 1 - 2 | 219 | 76 | ||
AK079556.1 | - | 1 - 2 | 139 | 76 | ||
AK080238.1 | - | 1 - 2 | 184 | 76 | ||
AK161465.1 | - | 1 - 2 | 75 | |||
AK165398.1 | - | 1 - 2 | 304 | 76 | ||
BC031761.1 | - | 1 - 2 | 168 | 4235549 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |