Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177034.3 | NP_796008.2 | 6 - 7 | 1818 | 72 | |
GenBank mRNA | AK032261.1 | - | 5 - 6 | 582 | 72 | |
AK147583.1 | - | 6 - 7 | 1808 | 72 | ||
AK147623.1 | - | 6 - 7 | 1770 | 72 | ||
AK188637.1 | - | 2 - 3 | 172 | 72 | ||
BC141181.1 | - | 6 - 7 | 1636 | 9055793 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |