Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339717 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339719 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339720 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339721 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P521052 |
BC049723.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P521052 |
NM_029627.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P521052 |
AK010485.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P521052 |
BC049723.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P521052 |
NM_029627.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P521052 |
AK010485.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489459 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379462 |
L05670.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379462 |
NM_013492.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379462 |
XM_001475611.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
295017 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817953 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
260136 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
691450 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
54231 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
263815 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
10448 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
509793 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
367056 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
179274 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P138852 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P521052 |
BC049723.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P521052 |
NM_029627.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P521052 |
AK010485.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P521052 |
BC049723.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P521052 |
NM_029627.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P521052 |
AK010485.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339732 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339740 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339705 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339746 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339706 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339744 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339717 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339735 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339730 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339709 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339707 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339742 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339733 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339727 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339714 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339748 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339722 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339738 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339710 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339713 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339712 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339716 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339719 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339702 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339720 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30339704 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08418331 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P138852 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08418337 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03071636 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P521052 |
BC049723.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P521052 |
NM_029627.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P521052 |
AK010485.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P521052 |
BC049723.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P521052 |
NM_029627.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P521052 |
AK010485.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P101410 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P138852 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08418331 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12358004 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12177086 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12358003 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12358006 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12358002 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12358000 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339717 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339721 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339719 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30339720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30339735 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12040016 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12155457 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12124798 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339640 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339716 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12175886 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12390995 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12374707 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339681 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339752 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339737 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339706 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339645 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339732 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339689 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339717 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339677 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339714 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339696 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339695 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11836673 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339713 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339638 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12374706 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339757 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339637 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339722 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339749 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339755 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12439019 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339629 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11867494 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339693 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339703 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12248295 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339744 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339697 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339632 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339621 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12175888 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12358006 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12357491 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339685 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339687 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339731 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12155432 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11836667 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339647 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339702 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339700 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339756 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339619 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339709 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339683 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339712 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12357493 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339727 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339622 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339746 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339748 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339679 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11867495 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12177087 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339707 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339624 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339705 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12222622 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339729 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339686 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339704 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30339710 |
- |
- |