Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289693.1 | NP_001276622.1 | 3 - 4 | 916 | 66 | |
GenBank mRNA | AK042348.1 | - | 3 - 4 | 918 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |