Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174857.3 | NP_777282.1 | 11 - 12 | 1911 | 65 | |
GenBank mRNA | AK004794.1 | - | 10 - 11 | 1774 | 65 | |
AK035566.1 | - | 11 - 12 | 1912 | 65 | ||
AK154219.1 | - | 11 - 12 | 1988 | 65 | ||
BC042773.1 | - | 11 - 12 | 1940 | 4019075 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |