Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090055.1 | - | 4 - 5 | 432 | 65 | |
AK166389.1 | - | 4 - 5 | 386 | 65 | ||
BC141167.1 | - | 4 - 5 | 368 | 9055779 | 65 | |
CT010280.1 | - | 2 - 3 | 197 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |