Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023134.4 | NP_075623.2 | 2 - 3 | 78 | 90 | |
GenBank mRNA | AK004620.1 | - | 2 - 3 | 79 | 90 | |
AK004788.1 | - | 2 - 3 | 79 | 90 | ||
BC061096.1 | - | 2 - 3 | 78 | 30300708 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |