Search Thermo Fisher Scientific
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026792.3 | NP_081068.1 | 6 - 7 | 1030 | 82 | |
GenBank mRNA | AK003649.1 | - | 6 - 7 | 774 | 82 | |
AK082137.1 | - | 6 - 7 | 805 | 82 | ||
AK089885.1 | - | 6 - 7 | 833 | 82 | ||
AK152709.1 | - | 6 - 7 | 773 | 82 | ||
AK152889.1 | - | 6 - 7 | 777 | 82 | ||
AK155378.1 | - | 7 - 8 | 979 | 82 | ||
AK170093.1 | - | 6 - 7 | 774 | 82 | ||
AY161042.1 | - | 6 - 7 | 1007 | 82 | ||
BC031987.1 | - | 6 - 7 | 755 | 3982362 | 82 | |
BC062903.1 | - | 6 - 7 | 1030 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |