Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011960.2 | NP_036090.2 | 4 - 5 | 1710 | 125 | |
GenBank mRNA | AF079557.1 | - | 4 - 5 | 1459 | 125 | |
AK036656.1 | - | 4 - 5 | 1733 | 125 | ||
AK088708.1 | - | 1 - 2 | 65 | 125 | ||
AK160742.1 | - | 3 - 4 | 1184 | 125 | ||
AK164013.1 | - | 4 - 5 | 1697 | 125 | ||
BC050892.1 | - | 4 - 5 | 1709 | 125 | ||
BC053037.1 | - | 4 - 5 | 1693 | 125 | ||
BC059827.1 | - | 4 - 5 | 1719 | 125 | ||
EF382673.1 | - | 2 - 3 | 419 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |