Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Corin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122756.1 | NP_001116228.1 | 10 - 11 | 1462 | 86 | |
NM_016869.3 | NP_058565.2 | 10 - 11 | 1619 | 86 | ||
GenBank mRNA | AB013874.1 | - | 10 - 11 | 1619 | 86 | |
BC093485.1 | - | 10 - 11 | 1395 | 30648458 | 86 | |
BC138339.1 | - | 10 - 11 | 1602 | 8861359 | 86 | |
BC138340.1 | - | 10 - 11 | 1602 | 8861360 | 86 | |
BC145243.1 | - | 10 - 11 | 1602 | 9053795 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |