Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF169300.1 | - | 1 - 2 | 119 | 59 | |
AK019171.1 | - | 1 - 2 | 134 | 59 | ||
AK133924.1 | - | 1 - 2 | 219 | 59 | ||
AK145596.1 | - | 1 - 2 | 96 | 59 | ||
AK151116.1 | - | 1 - 2 | 127 | 59 | ||
AK158987.1 | - | 1 - 2 | 136 | 59 | ||
AK159296.1 | - | 1 - 2 | 134 | 59 | ||
AK159778.1 | - | 1 - 2 | 128 | 59 | ||
AK207872.1 | - | 1 - 2 | 57 | 59 | ||
AK219295.1 | - | 1 - 2 | 57 | 59 | ||
BC003282.1 | - | 1 - 2 | 98 | 3582133 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |