Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Calb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009788.4 | NP_033918.1 | 2 - 3 | 295 | 152 | |
GenBank mRNA | AK002635.1 | - | 2 - 3 | 297 | 152 | |
AK005081.1 | - | 2 - 3 | 294 | 152 | ||
AK005243.1 | - | 2 - 3 | 295 | 152 | ||
AK028050.1 | - | 2 - 3 | 296 | 152 | ||
AK028327.1 | - | 2 - 3 | 294 | 152 | ||
AK038856.1 | - | 296 | 152 | |||
AK048517.1 | - | 2 - 3 | 296 | 152 | ||
AK077499.1 | - | 2 - 3 | 294 | 152 | ||
BC016421.1 | - | 2 - 3 | 226 | 4527000 | 152 | |
M21531.1 | - | 2 - 3 | 275 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |