Save up to 31% on select cell culture essentials

Bundle and save

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm01219775_m1
            See other STAT3 GE Assays ›
            Gene Symbol:
            Stat3
            Gene Name:
            signal transducer and activator of transcription 3
            Gene Aliases:
            1110034C02Rik, AW109958, Aprf
            Chromosome Location:
            Chr.11: 100886810 - 100939570 on Build GRCm38
            Species:
            Mouse
            Species Specific ID (Flybase ID):
            -
            Assay ID Mm01219775_m1
            Best Coverage
            Size
            Availability Inventoried
            Catalog # 4331182
            Price (USD) 272.00
            Your Price
            Online offer (USD):256.65
            256.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Gene Symbol: Stat3
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_011486.4 NP_035616.1 8 - 9 1071 75
            NM_213659.2 NP_998824.1 8 - 9 1071 75
            NM_213660.2 NP_998825.1 8 - 9 1071 75
            GenBank mRNA AK145486.1 - 8 - 9 927 75
            AK153005.1 - 8 - 9 983 75
            AK153472.1 - 8 - 9 983 75
            AK197148.1 - 1 - 2 57 75
            AY299489.1 - 7 - 8 798 75
            AY299490.1 - 7 - 8 798 75
            BC003806.1 - 8 - 9 959 3593588 75
            BC019168.1 - 8 - 9 975 4923137 75
            L29278.1 - 8 - 9 1050 75
            U06922.1 - 8 - 9 860 75
            U08378.1 - 8 - 9 938 75
            U30709.1 - 8 - 9 964 75

            Back To Top

            More Information


            Updated Mapping Information

            This assay targets the same location as MTA transcript cluster(s) TC1100003790.mm.1

            Set Membership:

            Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

            Matching Ambion siRNAs:

            Click here to find siRNAs for Gene Stat3

            Panther Classification:

            Molecular Function -

            DNA-binding transcription factor

            Gene Ontology Categories:

            Function(s) Process(es)

            temperature homeostasis
            eye photoreceptor cell differentiation
            transcription, DNA-templated
            regulation of transcription, DNA-templated
            transcription from RNA polymerase II promoter
            protein import into nucleus
            acute-phase response
            signal transduction
            JAK-STAT cascade
            cell proliferation
            negative regulation of cell proliferation
            negative regulation of hydrogen peroxide biosynthetic process
            phosphorylation
            cytokine-mediated signaling pathway
            stem cell population maintenance
            sexual reproduction
            intracellular receptor signaling pathway
            response to estradiol
            cellular response to hormone stimulus
            response to cytokine
            regulation of multicellular organism growth
            glucose homeostasis
            eating behavior
            negative regulation of apoptotic process
            positive regulation of Notch signaling pathway
            negative regulation of glycolytic process
            positive regulation of transcription, DNA-templated
            positive regulation of transcription from RNA polymerase II promoter
            regulation of mitochondrial membrane permeability
            astrocyte differentiation
            radial glial cell differentiation
            growth hormone receptor signaling pathway
            JAK-STAT cascade involved in growth hormone signaling pathway
            negative regulation of cell death
            interleukin-6-mediated signaling pathway
            negative regulation of neuron death
            positive regulation of growth factor dependent skeletal muscle satellite cell proliferation
            positive regulation of ATP biosynthetic process
            negative regulation of neuron migration
            RNA polymerase II core promoter proximal region sequence-specific DNA binding
            RNA polymerase II transcription factor activity, sequence-specific DNA binding
            RNA polymerase II repressing transcription factor binding
            transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
            DNA binding
            transcription factor activity, sequence-specific DNA binding
            signal transducer activity
            RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
            protein binding
            transcription factor binding
            protein kinase binding
            protein phosphatase binding
            CCR5 chemokine receptor binding
            glucocorticoid receptor binding
            sequence-specific DNA binding
            transcription regulatory region DNA binding
            protein dimerization activity

            MicroArray Probes:

            Affymetrix

            Array Name Probe IDs Targets Target Type
            Affymetrix Mouse Clariom S Array PSR1100034982.mm.1 NM_011486.5 Refseq
            Affymetrix Mouse Clariom S Array PSR1100034982.mm.1 NM_213659.3 Refseq
            Affymetrix Mouse Clariom S Array PSR1100034982.mm.1 NM_213660.3 Refseq
            Affymetrix Mouse Clariom S Array PSR1100034982.mm.1 TC1100003790.mm.1 MTA TC

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-kfh2r:80/100.66.79.247:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0