Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290383.1 | NP_001277312.1 | 7 - 8 | 1095 | 97 | |
NM_194055.3 | NP_918944.2 | 7 - 8 | 1098 | 97 | ||
GenBank mRNA | AK042164.1 | - | 7 - 8 | 710 | 97 | |
AK140848.1 | - | 7 - 8 | 1096 | 97 | ||
BC031468.1 | - | 5 - 6 | 401 | 4037840 | 97 | |
BC059280.1 | - | 5 - 6 | 399 | 97 | ||
DV654302.1 | - | 2 - 3 | 159 | 97 | ||
DV659240.1 | - | 5 - 6 | 454 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |