Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194344.2 | NP_919325.2 | 11 - 12 | 1670 | 77 | |
GenBank mRNA | AK028482.1 | - | 7 - 8 | 1766 | 77 | |
AK131143.1 | - | 5 - 6 | 535 | 77 | ||
AK150064.1 | - | 11 - 12 | 1448 | 77 | ||
AK155330.1 | - | 11 - 12 | 1432 | 77 | ||
AK159089.1 | - | 11 - 12 | 1449 | 77 | ||
AK170383.1 | - | 11 - 12 | 1503 | 77 | ||
AK170396.1 | - | 11 - 12 | 1450 | 77 | ||
AK171209.1 | - | 11 - 12 | 1427 | 77 | ||
AK171242.1 | - | 11 - 12 | 1427 | 77 | ||
BC024909.1 | - | 9 - 10 | 1120 | 77 | ||
BC056461.1 | - | 9 - 10 | 1014 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |