Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029578.3 | NP_083854.3 | 4 - 5 | 347 | 96 | |
GenBank mRNA | AK011555.1 | - | 4 - 5 | 338 | 96 | |
AK154341.1 | - | 4 - 5 | 324 | 96 | ||
AK165955.1 | - | 4 - 5 | 331 | 96 | ||
BY710811.1 | - | 4 - 5 | 335 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |