Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197028.1 | NP_001183957.1 | 3 - 4 | 487 | 77 | |
NM_028813.2 | NP_083089.1 | 4 - 5 | 553 | 77 | ||
GenBank mRNA | AF454755.1 | - | 4 - 5 | 470 | 77 | |
AK013193.1 | - | 4 - 5 | 464 | 77 | ||
AK051606.1 | - | 3 - 4 | 378 | 77 | ||
BC034120.1 | - | 4 - 5 | 476 | 4527027 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |