Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013814.1 | NP_001013836.1 | 4 - 5 | 485 | 59 | |
GenBank mRNA | BC132277.1 | - | 4 - 5 | 537 | 40130554 | 59 |
BC132279.1 | - | 4 - 5 | 537 | 40130556 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |