Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011215.2 | NP_035345.2 | 21 - 22 | 3003 | 67 | |
GenBank mRNA | AK138655.1 | - | 21 - 22 | 3092 | 67 | |
BC133678.1 | - | 21 - 22 | 2887 | 40109882 | 67 | |
U57345.1 | - | 21 - 22 | 2890 | 67 | ||
U82439.1 | - | 16 - 17 | 2011 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |