Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008243.3 | NP_032269.3 | 7 - 8 | 846 | 84 | |
GenBank mRNA | AK040934.1 | - | 7 - 8 | 830 | 84 | |
AK134222.1 | - | 7 - 8 | 829 | 84 | ||
AK144824.1 | - | 7 - 8 | 809 | 84 | ||
BC010551.1 | - | 7 - 8 | 831 | 4195560 | 84 | |
BC051393.1 | - | 7 - 8 | 815 | 5055055 | 84 | |
M74181.1 | - | 7 - 8 | 803 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |