Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167799.1 | NP_001161271.1 | 6 - 7 | 714 | 105 | |
NM_019829.4 | NP_062803.4 | 5 - 6 | 1013 | 105 | ||
GenBank mRNA | AK002722.1 | - | 6 - 7 | 696 | 105 | |
AK166087.1 | - | 5 - 6 | 1016 | 105 | ||
BC004849.1 | - | 6 - 7 | 647 | 2655845 | 105 | |
BC021883.1 | - | 5 - 6 | 996 | 3487476 | 105 | |
CN539678.1 | - | 6 - 7 | 678 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |