Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161413.1 | NP_001154885.1 | 7 - 8 | 1371 | 79 | |
NM_008577.4 | NP_032603.3 | 6 - 7 | 1130 | 79 | ||
GenBank mRNA | AB023408.1 | - | 6 - 7 | 1085 | 79 | |
AK090046.1 | - | 6 - 7 | 1108 | 79 | ||
AK156285.1 | - | 6 - 7 | 602 | 79 | ||
AK161280.1 | - | 7 - 8 | 1372 | 79 | ||
AK165417.1 | - | 6 - 7 | 1112 | 79 | ||
AK165550.1 | - | 6 - 7 | 1108 | 79 | ||
AK206213.1 | - | 1 - 2 | 71 | 79 | ||
BC065173.1 | - | 6 - 7 | 1090 | 6415269 | 79 | |
X14309.1 | - | 6 - 7 | 1072 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |