Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009205.2 | NP_033231.2 | 4 - 5 | 968 | 135 | |
GenBank mRNA | AK052623.1 | - | 4 - 5 | 939 | 135 | |
AK132275.1 | - | 4 - 5 | 940 | 135 | ||
BC013441.1 | - | 4 - 5 | 910 | 4222972 | 135 | |
D88533.1 | - | 4 - 5 | 928 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |