Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033289.2 | NP_001028461.2 | 1 - 2 | 449 | 81 | |
GenBank mRNA | AK077026.1 | - | 1 - 2 | 541 | 81 | |
AK138779.1 | - | 1 - 2 | 826 | 81 | ||
AK166114.1 | - | 1 - 2 | 565 | 81 | ||
BC103685.1 | - | 1 - 2 | 449 | 40048593 | 81 | |
BC104737.1 | - | 1 - 2 | 449 | 40048591 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |