Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291152.1 | NP_001278081.1 | 6 - 7 | 632 | 85 | |
NM_001291154.1 | NP_001278083.1 | 7 - 8 | 1351 | 85 | ||
NM_013731.3 | NP_038759.1 | 7 - 8 | 750 | 85 | ||
GenBank mRNA | AF169033.1 | - | 7 - 8 | 685 | 85 | |
AK050009.1 | - | 7 - 8 | 1332 | 85 | ||
AK136559.1 | - | 7 - 8 | 702 | 85 | ||
BC026549.1 | - | 5 - 6 | 377 | 4973908 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |