Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163504.1 | NP_001156976.1 | 8 - 9 | 1361 | 112 | |
NM_001163700.1 | NP_001157172.1 | 7 - 8 | 1157 | 112 | ||
NM_009108.2 | NP_033134.2 | 7 - 8 | 1145 | 112 | ||
GenBank mRNA | AK002513.1 | - | 7 - 8 | 1143 | 112 | |
AK132255.1 | - | 7 - 8 | 1137 | 112 | ||
BC015261.1 | - | 7 - 8 | 1116 | 4241705 | 112 | |
U09416.1 | - | 7 - 8 | 1114 | 112 | ||
U09417.1 | - | 8 - 9 | 1325 | 112 | ||
U09418.1 | - | 8 - 9 | 1322 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |