Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Git1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004144.1 | NP_001004144.1 | 15 - 16 | 1868 | 53 | |
GenBank mRNA | AK050701.1 | - | 2 - 3 | 186 | 53 | |
BC079870.1 | - | 15 - 16 | 1868 | 6831910 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |