Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159551.1 | NP_001153023.1 | 3 - 4 | 491 | 64 | |
NM_001159552.1 | NP_001153024.1 | 3 - 4 | 491 | 64 | ||
NM_010376.4 | NP_034506.1 | 3 - 4 | 491 | 64 | ||
GenBank mRNA | AF483214.1 | - | 3 - 4 | 458 | 64 | |
AF515662.1 | - | 3 - 4 | 370 | 64 | ||
AJ345032.1 | - | 3 - 4 | 501 | 64 | ||
AK007664.1 | - | 3 - 4 | 473 | 64 | ||
AK012600.1 | - | 3 - 4 | 484 | 64 | ||
AK028109.1 | - | 3 - 4 | 460 | 64 | ||
AK039008.1 | - | 3 - 4 | 465 | 64 | ||
AK049101.1 | - | 3 - 4 | 465 | 64 | ||
AK050664.1 | - | 3 - 4 | 434 | 64 | ||
AK080966.1 | - | 3 - 4 | 453 | 64 | ||
AK159176.1 | - | 3 - 4 | 465 | 64 | ||
AK169922.1 | - | 3 - 4 | 448 | 64 | ||
AK171790.1 | - | 3 - 4 | 419 | 64 | ||
BC056977.1 | - | 3 - 4 | 490 | 6818912 | 64 | |
CA321999.1 | - | 3 - 4 | 491 | 64 | ||
DQ168449.1 | - | 3 - 4 | 507 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |