Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144892.1 | NP_659141.1 | 3 - 4 | 488 | 102 | |
GenBank mRNA | BC017152.1 | - | 3 - 4 | 488 | 4511696 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |