Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018771.3 | NP_061241.1 | 3 - 4 | 572 | 85 | |
GenBank mRNA | AF061263.1 | - | 2 - 3 | 506 | 85 | |
AF104358.1 | - | 3 - 4 | 550 | 85 | ||
AK002492.1 | - | 3 - 4 | 553 | 85 | ||
AK086506.1 | - | 3 - 4 | 573 | 85 | ||
BC003490.1 | - | 3 - 4 | 572 | 2811749 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |