Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172945.2 | NP_766533.2 | 7 - 8 | 943 | 74 | |
GenBank mRNA | BC056928.1 | - | 2 - 3 | 172 | 74 | |
BC151158.1 | - | 7 - 8 | 921 | 9088046 | 74 | |
BC172028.1 | - | 7 - 8 | 922 | 74 | ||
CA315262.1 | - | 6 - 7 | 629 | 74 | ||
CD578820.1 | - | 1 - 2 | 41 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |