Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081241.2 | NP_001074710.2 | 10 - 11 | 952 | 87 | |
GenBank mRNA | AK052316.1 | - | 10 - 11 | 901 | 87 | |
AK122573.1 | - | 10 - 11 | 926 | 87 | ||
BC079880.1 | - | 10 - 11 | 866 | 6852251 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |