Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB480706.1 | - | 4 - 5 | 814 | 88 | |
AK138730.1 | - | 6 - 7 | 794 | 88 | ||
AK151547.1 | - | 6 - 7 | 961 | 88 | ||
AK156667.1 | - | 6 - 7 | 957 | 88 | ||
AK160328.1 | - | 6 - 7 | 957 | 88 | ||
AK171877.1 | - | 6 - 7 | 953 | 88 | ||
BC061255.1 | - | 6 - 7 | 960 | 30251263 | 88 | |
U59463.1 | - | 6 - 7 | 946 | 88 | ||
Y13089.1 | - | 6 - 7 | 938 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |