Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083319.1 | NP_001076788.1 | 13 - 14 | 1679 | 81 | |
NM_013699.2 | NP_038727.1 | 12 - 13 | 1571 | 81 | ||
GenBank mRNA | AB221581.1 | - | 11 - 12 | 1159 | 81 | |
AK079815.1 | - | 5 - 6 | 406 | 81 | ||
AK140942.1 | - | 7 - 8 | 576 | 81 | ||
AK146040.1 | - | 1 - 2 | 191 | 81 | ||
AY182062.1 | - | 12 - 13 | 1268 | 81 | ||
BC034749.1 | - | 11 - 12 | 1898 | 81 | ||
BC139828.1 | - | 11 - 12 | 1188 | 40142981 | 81 | |
BC141082.1 | - | 11 - 12 | 1250 | 9055694 | 81 | |
U20086.1 | - | 11 - 12 | 1162 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |