Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144829.1 | NP_659078.1 | 1 - 2 | 81 | 78 | |
GenBank mRNA | AK078016.1 | - | 1 - 2 | 50 | 78 | |
AK136332.1 | - | 1 - 2 | 48 | 78 | ||
AK168281.1 | - | 1 - 2 | 48 | 78 | ||
BC005711.1 | - | 1 - 2 | 81 | 3497962 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |