Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019835.2 | NP_062809.2 | 4 - 5 | 634 | 80 | |
GenBank mRNA | AB004786.1 | - | 4 - 5 | 635 | 80 | |
AK151469.1 | - | 1 - 2 | 104 | 80 | ||
AK152652.1 | - | 1 - 2 | 104 | 80 | ||
BE981379.1 | - | 4 - 5 | 504 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |