Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039242.2 | NP_001034331.1 | 3 - 4 | 447 | 95 | |
NM_172568.2 | NP_766156.2 | 3 - 4 | 447 | 95 | ||
GenBank mRNA | AK132734.1 | - | 3 - 4 | 436 | 95 | |
BC100474.1 | - | 3 - 4 | 440 | 30918723 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |