Search Thermo Fisher Scientific
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153065.3 | NP_694705.2 | 9 - 10 | 1045 | 78 | |
GenBank mRNA | AK134274.1 | - | 9 - 10 | 1046 | 78 | |
AK138442.1 | - | 9 - 10 | 1069 | 78 | ||
AK146469.1 | - | 9 - 10 | 1073 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |