Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009764.3 | NP_033894.3 | 20 - 21 | 5369 | 67 | |
GenBank mRNA | AB221610.1 | - | 11 - 12 | 2598 | 67 | |
BC068303.1 | - | 20 - 21 | 5202 | 30431022 | 67 | |
U31625.1 | - | 20 - 21 | 5226 | 67 | ||
U32446.1 | - | 20 - 21 | 5279 | 67 | ||
U36475.1 | - | 19 - 20 | 5189 | 67 | ||
U68174.1 | - | 20 - 21 | 5273 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |