Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193645.1 | NP_001180574.1 | 7 - 8 | 905 | 83 | |
NM_016722.4 | NP_057931.3 | 7 - 8 | 905 | 83 | ||
GenBank mRNA | AF111346.1 | - | 7 - 8 | 826 | 83 | |
AK033946.1 | - | 7 - 8 | 823 | 83 | ||
AK159592.1 | - | 7 - 8 | 882 | 83 | ||
AK220245.1 | - | 7 - 8 | 807 | 83 | ||
CB600418.1 | - | 4 - 5 | 366 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |