Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025894.2 | NP_080170.1 | 7 - 8 | 845 | 72 | |
GenBank mRNA | AK005114.1 | - | 7 - 8 | 879 | 72 | |
AK007619.1 | - | 7 - 8 | 890 | 72 | ||
AK007680.1 | - | 7 - 8 | 819 | 72 | ||
AK009969.1 | - | 7 - 8 | 848 | 72 | ||
AK017843.1 | - | 7 - 8 | 862 | 72 | ||
AK150910.1 | - | 7 - 8 | 868 | 72 | ||
AK151242.1 | - | 7 - 8 | 846 | 72 | ||
AK152041.1 | - | 7 - 8 | 868 | 72 | ||
AK152240.1 | - | 7 - 8 | 877 | 72 | ||
AK160283.1 | - | 7 - 8 | 867 | 72 | ||
AK162779.1 | - | 846 | 72 | |||
AK167236.1 | - | 7 - 8 | 853 | 72 | ||
BC004694.1 | - | 7 - 8 | 863 | 3498952 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |