Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183023.1 | NP_898844.1 | 1 - 2 | 96 | 67 | |
GenBank mRNA | AK134613.1 | - | 1 - 2 | 342 | 67 | |
AY326955.1 | - | 1 - 2 | 96 | 67 | ||
BC115475.1 | - | 1 - 2 | 95 | 67 | ||
BC115476.1 | - | 5 - 6 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |