Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139272.2 | NP_644678.2 | 13 - 14 | 1445 | 69 | |
GenBank mRNA | AF348968.1 | - | 13 - 14 | 1313 | 69 | |
AK046567.1 | - | 13 - 14 | 1403 | 69 | ||
AK145122.1 | - | 13 - 14 | 1323 | 69 | ||
AK149609.1 | - | 13 - 14 | 1373 | 69 | ||
AK151536.1 | - | 13 - 14 | 1236 | 69 | ||
BC007172.1 | - | 12 - 13 | 1180 | 69 | ||
BC053063.1 | - | 12 - 13 | 1288 | 5702572 | 69 | |
BC059818.1 | - | 13 - 14 | 1390 | 5697789 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |