Search Thermo Fisher Scientific
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019840.2 | NP_062814.2 | 5 - 6 | 872 | 62 | |
GenBank mRNA | AF326555.1 | - | 5 - 6 | 612 | 62 | |
AF326556.1 | - | 5 - 6 | 612 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |