Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rqcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021383.5 | NP_067358.1 | 925 | 86 | ||
GenBank mRNA | AF221849.1 | - | 5 - 6 | 547 | 86 | |
AK005025.2 | - | 5 - 6 | 927 | 86 | ||
BC050898.1 | - | 5 - 6 | 726 | 6401907 | 86 | |
BC051948.1 | - | 764 | 5708192 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |