Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172695.2 | NP_766283.2 | 7 - 8 | 1255 | 78 | |
GenBank mRNA | AK019070.1 | - | 7 - 8 | 1261 | 78 | |
AK075934.1 | - | 7 - 8 | 1187 | 78 | ||
AK166122.1 | - | 7 - 8 | 1161 | 78 | ||
BC139355.1 | - | 7 - 8 | 1254 | 8862377 | 78 | |
BC139356.1 | - | 7 - 8 | 1254 | 8862378 | 78 | |
BC139773.1 | - | 7 - 8 | 1165 | 40111789 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |