Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026295.2 | NP_080571.2 | 4 - 5 | 746 | 53 | |
GenBank mRNA | AK156617.1 | - | 4 - 5 | 775 | 53 | |
BC053435.1 | - | 4 - 5 | 746 | 6394578 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |