Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293728.1 | NP_001280657.1 | 8 - 9 | 837 | 70 | |
NM_011656.3 | NP_035786.1 | 9 - 10 | 912 | 70 | ||
GenBank mRNA | AF002860.1 | - | 9 - 10 | 806 | 70 | |
AF047704.1 | - | 9 - 10 | 846 | 70 | ||
AK132488.1 | - | 9 - 10 | 844 | 70 | ||
AK171705.1 | - | 8 - 9 | 802 | 70 | ||
BC019213.1 | - | 9 - 10 | 843 | 5064658 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |